(1S)-(+)-Camphor-10-sulphonyl chloride ≥90%

Supplier: Ambeed
Danger

Synonyms: (1S)-(+)-2-Oxobornane-10-sulphonyl chloride, ((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonyl chloride, D(+)-Camphor-10-sulphonyl chloride

A1003443-25G A1003443-5G
77570-022EA 58 USD
77570-022 77782-717
(1S)-(+)-Camphor-10-sulphonyl chloride ≥90%
(1S)-(+)-Camphor-10-sulphonyl chloride

Precaución: for Research Use Only

Formula: C₁₀H₁₅ClO₃S
MDL Number: MFCD00064156
CAS Number: 21286-54-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR