2-Iodo-4-methoxyaniline ≥95%

Supplier: Ambeed
Warning

A1003409-1G A1003409-5G A1003409-25G A1003409-100G
77592-052EA 34.85 USD
77592-052 77595-650 77619-504 77782-716
2-Iodo-4-methoxyaniline ≥95%
2-Iodo-4-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₈INO
MDL Number: MFCD07776739
CAS Number: 191348-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR