Bismuth(III) chloride ≥97% (dry basis) may cont. up to 3% water

Supplier: Ambeed
Warning

Synonyms: Bismuth(III) chloride, Bismuth trichloride

A1002938-100G A1002938-500G A1002938-1000G
77592-048EA 48.18 USD
77592-048 77595-646 77782-710
Bismuth(III) chloride ≥97% (dry basis) may cont. up to 3% water
Bismuth(III) chloride

Precaución: for Research Use Only

Formula: BiCl₃
MW: 315.34 g/mol
Boiling Pt: 447
Melting Pt: 232
MDL Number: MFCD00003461
CAS Number: 7787-60-2
Merck Index: 13,01261

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR