V-9302 ≥99%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-4-(bis(2-((3-methylbenzyl)oxy)benzyl)amino)butanoic acid

A1002473-10MG A1002473-5MG
77595-640EA 128.13 USD
77595-640 77595-642
V-9302 ≥99%
V-9302

Precaución: for Research Use Only

Formula: C₃₄H₃₈N₂O₄
MW: 538.69 g/mol
MDL Number: MFCD32062750
CAS Number: 1855871-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR