4-(1-Hydroxypropyl)benzotrifluoride ≥95%

Supplier: Ambeed
Warning

Synonyms: 1-[4-(Trifluoromethyl)phenyl]propanol, 1-(4-(Trifluoromethyl)phenyl)propan-1-ol

A100189-250MG A100189-5G
77682-927EA 20 USD
77682-927 77682-928
4-(1-Hydroxypropyl)benzotrifluoride ≥95%
4-(1-Hydroxypropyl)benzotrifluoride

Precaución: For research use only.

Formula: C₁₀H₁₁F₃O
MW: 204.19 g/mol
MDL Number: MFCD07773046
CAS Number: 67081-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR