(R)-tert-Butyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A1001555-250MG A1001555-5G
77569-998EA 106.09 USD
77569-998 77619-476
(R)-tert-Butyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate ≥98%
(R)-tert-Butyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₇F₂NO₃
MW: 237.2400 g/mol
CAS Number: 1893408-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR