Mal-amido-PEG2-NHS ≥95%

Supplier: Ambeed
Warning

Synonyms: Maleimide-PEG2-succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 3-(2-(2-(3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido)ethoxy)ethoxy)propanoate, N-Succinimidyl 13-Maleimido-11-oxo-4,7-dioxa-10-azatridecanoate, 13-Maleimido-11-oxo-4,7-dioxa-10-azatridecanoic acid N-Succinimidyl Ester, Maleimide-PEG2-NHS ester, 2,5-Doxopyrrolidin-1-yl 3-[2-[2-[3-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanamido]ethoxy]ethoxy]propanoate

A100037-100MG A100037-5G A100037-1G A100037-250MG
77610-722EA 19.57 USD
77610-722 77610-724 77682-924 77682-925
Mal-amido-PEG2-NHS ≥95%
Mal-amido-PEG2-NHS

Precaución: For research use only.

Formula: C₁₈H₂₃N₃O₉
MW: 425.39 g/mol
MDL Number: MFCD11041137
CAS Number: 955094-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR