Zn(II) Phthalocyanine tetrasulfonic acid ≥98%

Supplier: Ambeed
Warning

A1000214-100MG A1000214-250MG A1000214-50MG A1000214-1G
77619-458EA 270.89 USD
77619-458 77619-460 77619-462 77782-675
Zn(II) Phthalocyanine tetrasulfonic acid ≥98%
Zn(II) Phthalocyanine tetrasulfonic acid

Precaución: for Research Use Only

Formula: C₃₂H₁₆N₈O₁₂S₄Zn
MW: 898.1848 g/mol
MDL Number: MFCD00672308
CAS Number: 61586-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR