Brilliant green solution

Supplier: G-Biosciences

Synonyms: Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate

786-2111 786-2112
77540-280EA 29.85 USD
77540-280 77540-282
Brilliant green solution
Brilliant green

Brilliant Green is used as a counterstain.


  • Quick staining dye


It differentiates acid-fast bacteria from non-acid-fast bacteria

Formula: C₂₇H₃₄N₂O₄S
MW: 482.65 g/mol
MDL Number: MFCD00011880
CAS Number: 633-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR