CP-310™ Alkaline Process and Research Cleaner

Supplier: Steris
1K7401 1K7405 1K74D3
77531-108EA 3064.1 USD
77531-108 77531-112 77531-092
CP-310™ Alkaline Process and Research Cleaner
Cleaning Agents Detergents and Surface Cleansers
For use in CIP, COP and manual applications.

CP-310™ Alkaline detegent is formulated with a proprietary blend of low-mercury grade potassium hydroxide, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants, and is ideal for use in CIP, COP and manual applications.

CP-310™ Alkaline process and research cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

CAS: 1310-58-3; 64-02-8; 9003-01-4
UN 1814
Hazard class: 8
Packing group: II

Precaución: Danger, GHS07; GHS05.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR