Human Recombinant NRAS, Active (Biotinylated, His tag) (from E. coli), Biotinylated
Supplier: Sino Biological
R107-311HB-10 R107-311HB-50 R107-311HB-500
77563-076EA 398 USD
77563-076 77563-078 77563-080
Human Recombinant NRAS, Active (Biotinylated, His tag) (from E. coli), Biotinylated
Proteins and Peptides
Recombinant human NRAS (1-169) expressed in E. coli cells using an N-terminal Avi-His tag and enzymatically biotinylated in vitro prior to the final chromatography purification.
  • Shipping temperature: Dry ice