Human Recombinant HRAS, Active (Biotinylated, His tag) (from E. coli), Biotinylated
Supplier: Sino Biological
R105-310HB-10 R105-310HB-50 R105-310HB-500
77563-028EA 398 USD
77563-028 77563-030 77563-032
Human Recombinant HRAS, Active (Biotinylated, His tag) (from E. coli), Biotinylated
Proteins and Peptides
Recombinant human HRAS (2-186) expressed in E. coli cells using an N-terminal Avi-His tag and enzymatically biotinylated in vitro prior to the final chromatography purification.
  • Shipping temperature: Dry ice