Human Recombinant NRAS (Q61K, His tag), Biotinylated (from E. coli), Biotinylated
Supplier: Sino Biological
R07-32HB-100 R07-32HB-1000 R07-32HB-20
77562-940EA 1108 USD
77562-940 77562-942 77562-944
Human Recombinant NRAS (Q61K, His tag), Biotinylated (from E. coli), Biotinylated
Proteins and Peptides
Recombinant human NRAS (Q61K) (1-186) expressed in E. coli cells using an N-terminal Avi-His tag and enzymatically biotinylated in vitro prior to the final chromatography purification. NRAS (Q61K) is constitutively active.
  • Shipping temperature: Dry ice