Human Recombinant Parkin (GST tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P215-30G-100 P215-30G-1000 P215-30G-20
77561-310EA 688 USD
77561-310 77561-312 77561-314
Human Recombinant Parkin (GST tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human Parkin was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice