Human Recombinant PFTK1 (CDK14, GST tag)/CyclinY, Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P15-10G-50 P15-10G-500
77561-192EA 1168 USD
77561-192 77561-194
Human Recombinant PFTK1 (CDK14, GST tag)/CyclinY, Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human PFTK1 (CDK14) and Cyclin Y (2-end) were co-expressed by baculovirus in Sf9 insect cells using N-terminal GST tags.
  • Shipping temperature: Dry ice