Human Recombinant FLT3-ITD-W51 (GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
F12-12CG-50 F12-12CG-500 F12-12CG-10
77557-648EA 1158 USD
77557-648 77557-650 77557-646
Human Recombinant FLT3-ITD-W51 (GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human FLT3-ITD-W51 (571-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.