Human Recombinant FLT3 (D835Y, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
F12-12BG-10 F12-12BG-50 F12-12BG-500
77557-640EA 338 USD
77557-640 77557-642 77557-644
Human Recombinant FLT3 (D835Y, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human FLT3 (D835Y) (571-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.