Human Recombinant FGFR2 (E565A, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
F05-12KG-10 F05-12KG-50 F05-12KG-500
77557-476EA 338 USD
77557-476 77557-478 77557-480
Human Recombinant FGFR2 (E565A, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human FGFR2 (E565A) (285-end) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag.