Human Recombinant HER3 (His tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
E28-35H-100 E28-35H-1000 E28-35H-20
77557-204EA 818 USD
77557-204 77557-206 77557-208
Human Recombinant HER3 (His tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human HER3 (667-1020) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.