Human Recombinant HER2 (V777_G778insCG, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E27-132CG-10 E27-132CG-50 E27-132CG-500
77557-166EA 338 USD
77557-166 77557-168 77557-170
Human Recombinant HER2 (V777_G778insCG, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human HER2 (V777_G778insCG) (676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice