Human Recombinant HER2 (A775_G776insYVMA, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E27-132BG-10 E27-132BG-50 E27-132BG-500
77557-160EA 338 USD
77557-160 77557-162 77557-164
Human Recombinant HER2 (A775_G776insYVMA, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human HER2 (A775_G776insYVMA) (676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice