Human Recombinant HER2 (L755S L869R, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E27-12QG-10 E27-12QG-50 E27-12QG-500
77557-148EA 348 USD
77557-148 77557-150 77557-152
Human Recombinant HER2 (L755S L869R, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human HER2 (L755S L869R) (676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice