Human Recombinant HER2 (L755S, GST tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
E27-122EG-10 E27-122EG-50 E27-122EG-500
77557-082EA 338 USD
77557-082 77557-084 77557-086
Human Recombinant HER2 (L755S, GST tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human HER2 (L755S) (676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice