Human Recombinant HER2 (d755-759, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E27-122BG-10 E27-122BG-50 E27-122BG-500
77557-064EA 338 USD
77557-064 77557-066 77557-068
Human Recombinant HER2 (d755-759, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human HER2 (d755-759) (676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice