Human Recombinant HER2 (GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E27-11G-10 E27-11G-50 E27-11G-500
77557-058EA 348 USD
77557-058 77557-060 77557-062
Human Recombinant HER2 (GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human HER2 (676-end) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
  • Shipping temperature: Dry ice