Mouse Recombinant EPHA1 (GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E13-11G-10 E13-11G-50 E13-11G-500
77556-934EA 348 USD
77556-934 77556-936 77556-938
Mouse Recombinant EPHA1 (GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant mouse EPHA1 (569-end) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.