Human Recombinant EGFR (L792H, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E10-122J2G-10 E10-122J2G-50 E10-122J2G-500
77556-474EA 338 USD
77556-474 77556-476 77556-478
Human Recombinant EGFR (L792H, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human EGFR (L792H) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice