Human Recombinant EGFR (GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E10-112G-50 E10-112G-500
77556-332EA 1168 USD
77556-332 77556-334
Human Recombinant EGFR (GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human EGFR (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice