Human Recombinant P300, (GST Tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
P07-31G-100 P07-31G-1000 P07-31G-20
77561-010EA 1028 USD
77561-010 77561-012 77561-014
Human Recombinant P300, (GST Tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human P300 (1283 to 1673) was expressed by Baculovirus in Sf9 cells using a N to terminal GST tag.
  • Shipping temperature: Dry ice