Human Recombinant PAK1 (GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
P02B-10BG-10 P02B-10BG-50 P02B-10BG-500
77560-922EA 348 USD
77560-922 77560-924 77560-926
Human Recombinant PAK1 (GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length Human PAK1 was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.