Dog Recombinant NNMT (Dog) (GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
N330D-380G-10 N330D-380G-50 N330D-380G-500
77560-800EA 418 USD
77560-800 77560-802 77560-804
Dog Recombinant NNMT (Dog) (GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length Canis lupus NNMT was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.