Human Recombinant NF1 (His Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
N120-31H-10 N120-31H-50 N120-31H-500
77560-556EA 258 USD
77560-556 77560-558 77560-560
Human Recombinant NF1 (His Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human NF1 (1197 to 1528) was expressed in E. coli cells using an N to terminal His tag.