Human Recombinant MLCK (GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
M62-11G-10 M62-11G-50 M62-11G-500
77560-390EA 348 USD
77560-390 77560-392 77560-394
Human Recombinant MLCK (GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human MLCK (1425 to 1776) was expressed by Baculovirus in Sf9 insect cells using a N to terminal GST tag.
  • Shipping temperature: Dry ice