Human Recombinant MET (Del Ex14, GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
M52-12PG-10 M52-12PG-50 M52-12PG-500
77560-236EA 338 USD
77560-236 77560-238 77560-240
Human Recombinant MET (Del Ex14, GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human MET (Del Ex14) or (Del 963 to 1009) (956 to end) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.