Human Recombinant KAT2B (PCAF, GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
K311-381BG-50 K311-381BG-500 K311-381BG-10
77558-968EA 1298 USD
77558-968 77558-970 77558-966
Human Recombinant KAT2B (PCAF, GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human KAT2B (PCAF) (431 to end) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.