Human Recombinant INCENP, (His Tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
I30-31H-100 I30-31H-1000 I30-31H-20
77558-570EA 818 USD
77558-570 77558-572 77558-574
Human Recombinant INCENP, (His Tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human INCENP (783 to end) was expressed by Baculovirus in Sf9 insect cells using an N to terminal His tag. This product can be used for Aurora B and Aurora C activation in vitro.
  • Shipping temperature: Dry ice