Copper(I) 2-hydroxy-3-methylbenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Copper(I)-3-methylsalicylate

A121718-100G A121718-5G A121718-1G A121718-10G A121718-25G
77566-258EA 799.28 USD
77566-258 77566-260 77571-294 77611-034 77670-722
Copper(I) 2-hydroxy-3-methylbenzoate 95%
Copper(I) 2-hydroxy-3-methylbenzoate

Precaución: For research use only.

Formula: C₈H₇CuO₃
MW: 214.69 g/mol
MDL Number: MFCD10699180
CAS Number: 326477-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR