(1S,3R)-3-Aminocyclohexanecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: (1S,3R)-3-Aminocyclohexanecarboxylic acid

A994214-1G A994214-250MG A994214-100MG
77569-734EA 194.67 USD
77569-734 77614-716 77684-919
(1S,3R)-3-Aminocyclohexanecarboxylic acid 98%
(1S,3R)-3-Aminocyclohexanecarboxylic acid

Precaución: For research use only.

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
MDL Number: MFCD09753648
CAS Number: 81131-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR