(Triisopropylsiloxy)methyl chloride 85%

Supplier: Ambeed
Danger

A991979-1G A991979-250MG A991979-5G A991979-100MG
77569-726EA 90.64 USD
77569-726 77569-728 77573-982 77623-408
(Triisopropylsiloxy)methyl chloride 85%
(Triisopropylsiloxy)methyl chloride

Precaución: For research use only.

Formula: C₁₀H₂₃ClOSi
MW: 222.8275 g/mol
MDL Number: MFCD05865265
CAS Number: 217300-17-9
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR