tert-Butyl 3-methyl-4-oxopyrrolidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A989673-250MG A989673-1G A989673-5G
77569-718EA 44.29 USD
77569-718 77599-394 77623-364
tert-Butyl 3-methyl-4-oxopyrrolidine-1-carboxylate 98%
tert-Butyl 3-methyl-4-oxopyrrolidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
MDL Number: MFCD29918272
CAS Number: 885102-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR