3-Bromo-1-methyl-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

A974304-5G A974304-25G A974304-1G
77569-702EA 101.97 USD
77569-702 77573-924 77670-422
3-Bromo-1-methyl-1H-pyrrolo[2,3-b]pyridine 97%
3-Bromo-1-methyl-1H-pyrrolo[2,3-b]pyridine

Precaución: For research use only.

Formula: C₈H₇BrN₂
MW: 211.059 g/mol
MDL Number: MFCD09743454
CAS Number: 281192-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR