Methyl 6-chloro-2,4-dimethylnicotinate 95%

Supplier: Ambeed
Warning

A954103-1G A954103-250MG
77569-680EA 233.81 USD
77569-680 77684-896
Methyl 6-chloro-2,4-dimethylnicotinate 95%
Methyl 6-chloro-2,4-dimethylnicotinate

Precaución: For research use only.

Formula: C₉H₁₀ClNO₂
MW: 199.63 g/mol
MDL Number: MFCD18258696
CAS Number: 1256789-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR