Potassium (cyclopropylmethyl)trifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: potassium cyclopropylmethyl(trifluoro)boranuide

A948812-1G A948812-250MG
77569-670EA 152 USD
77569-670 77603-334
Potassium (cyclopropylmethyl)trifluoroborate 98%
Potassium (cyclopropylmethyl)trifluoroborate

Precaución: For research use only.

Formula: C₄H₇BF₃·K
MW: 162.003 g/mol
MDL Number: MFCD11052655
CAS Number: 1356481-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR