(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)methanol 97%

Supplier: Ambeed
Warning

A884197-250MG A884197-1G A884197-100MG
77569-612EA 22 USD
77569-612 77623-102 77784-312
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)methanol 97%
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)methanol

Precaución: For research use only.

Formula: C₈H₅BrF₄O
MDL Number: MFCD28401920
CAS Number: 1352718-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR