6-Methyl-3,6-diazabicyclo[3.1.1]heptane dihydrochloride 97%

Supplier: Ambeed
Warning

A872823-250MG A872823-1G A872823-100MG
77569-608EA 487 USD
77569-608 77784-289 77784-288
6-Methyl-3,6-diazabicyclo[3.1.1]heptane dihydrochloride 97%
6-Methyl-3,6-diazabicyclo[3.1.1]heptane dihydrochloride

Precaución: For research use only.

Formula: C₆H₁₄Cl₂N₂
MDL Number: MFCD31704972
CAS Number: 2231676-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR