2-[4-(Fluorosulfonyl)phenyl]acetic acid 95%

Supplier: Ambeed
Danger

A853948-100MG A853948-250MG
77569-580EA 108.15 USD
77569-580 77569-582
2-[4-(Fluorosulfonyl)phenyl]acetic acid 95%
2-[4-(Fluorosulfonyl)phenyl]acetic acid

Precaución: For research use only.

Formula: C₈H₇FO₄S
MW: 218.2 g/mol
MDL Number: MFCD24483006
CAS Number: 402-57-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR