5-Chloro-4-iodopyridin-2-amine 98%
Supplier: Ambeed
|
Warning
|
A837374-1G
A837374-100MG
A837374-250MG
77569-558EA
121.54
USD
77569-558
77614-504
77684-799
5-Chloro-4-iodopyridin-2-amine 98%
5-Chloro-4-iodopyridin-2-amine
Precaución: For research use only.
Formula:
C₅H₄ClIN₂ MW: 254.45 g/mol |
MDL Number:
MFCD18250085 CAS Number: 1260667-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...