tert-Butyl 3-(2-bromoethoxy)propanoate 97%

Supplier: Ambeed
Warning

Synonyms: Bromo-PEG1-C2-Boc

A835881-1G A835881-250MG A835881-100MG
77569-552EA 50.47 USD
77569-552 77569-554 77684-795
tert-Butyl 3-(2-bromoethoxy)propanoate 97%
tert-Butyl 3-(2-bromoethoxy)propanoate

Precaución: For research use only.

Formula: C₉H₁₇BrO₃
MW: 253.13 g/mol
MDL Number: MFCD22574803
CAS Number: 1393330-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR