β-Methylstyrene (cis- and trans- mixture) stabilised

Supplier: Ambeed
Danger

Synonyms: 1-Propenylbenzene, β-Methylstyrene, 1-Phenyl-1-propene

Stabilization: Stabilized with TBC

A833928-5G A833928-1G
77569-550EA 64 USD
77569-550 77900-992
β-Methylstyrene (cis- and trans- mixture) stabilised
beta-Methylstyrene (cis- and trans- mixture)

Precaución: For research use only.

Formula: C₉H₁₀
MDL Number: MFCD00009280
CAS Number: 637-50-3
UN: 3295
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR