2-[(tert-Butoxy)carbonyl]-2,3-dihydro-1H-isoindole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(tert-Butoxycarbonyl)isoindoline-5-carboxylic acid

A827333-1G A827333-100MG A827333-250MG
77569-542EA 252 USD
77569-542 77618-324 77747-584
2-[(tert-Butoxy)carbonyl]-2,3-dihydro-1H-isoindole-5-carboxylic acid 95%
2-[(tert-Butoxy)carbonyl]-2,3-dihydro-1H-isoindole-5-carboxylic acid

Precaución: For research use only.

Formula: C₁₄H₁₇NO₄
MDL Number: MFCD08234862
CAS Number: 149353-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR